High Opacity Paint Markers, Electron Microscopy Sciences
Product Details & Documents
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface
- Weatherproof
- Resistant to alcohol and acetone
- Contain xylene
Markers are available in black, red and white with bullet tips. Except model 130, it writes like a pen with white ink for marking on dark surfaces, including carbon tabs, which is use in SEM specimen mounting purpose.
Spécifications
- Désignation:Opaque White Pen