SUPERFROST® Microscope Slides, Frosted End, Electron Microscopy Sciences
SUPERFROST® Microscope Slides, Frosted End, Electron Microscopy Sciences
Superfrosted end slide, 1.0 mm
Catalog # 100493-524
Supplier #:  71867-10
SUPERFROST® Microscope Slides, Frosted End, Electron Microscopy Sciences
Catalog # 100493-524
SUPERFROST® Microscope Slides, Frosted End, Electron Microscopy Sciences
SUPERFROST® Microscope Slides, Frosted End, Electron Microscopy Sciences
Superfrosted end slide, 1.0 mm
Price
$1,063.15
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Microscope glass slides are precleaned and are made from clear-white glass. They are individually inspected and free from moisture, dirt, and film. All of our slides are flat, uniform, durable, and cellophane wrapped.
  • Made from the highest quality glass sheets
  • The dimensions of the slides have been standardized to millimeters
  • Packed with care to assure cleanliness
  • Glass is fully annealed to minimize chipping
Microscope glass slides are manufactured by Esco - Erie Scientific Co, where reliability and quality are recognized to be the best.
Specifications
  • Désignation:
    Superfrosted end slide
  • Épaisseur:
    1.0 mm