Replacement Desiccator Plates, Electron Microscopy Sciences
Replacement Desiccator Plates, Electron Microscopy Sciences
230 mm
Catalog # 100493-224
Supplier #:  71242-23
Replacement Desiccator Plates, Electron Microscopy Sciences
Catalog # 100493-224
Replacement Desiccator Plates, Electron Microscopy Sciences
Replacement Desiccator Plates, Electron Microscopy Sciences
230 mm
Price
$456.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These 4.8 mm thick plates are able to withstand temperatures up to 150°C (300°F) and have excellent resistance to thermal shock, and to chemicals. They are non-combustible and asbestos free. Round plates are perforated with 4 mm (0.165”) diameter holes on 12.7 mm (1/2”) centers in a straight line pattern. They are available in three sizes, and fit our desiccators.
Specifications
  • Taille:
    230 mm