Low Temperature Dessicator, Electron Microscopy Sciences | Avantor
Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Desiccator Cabinet, Bronze
Catalog # 100493-162
Fournisseur #:  71220-A
Low Temperature Dessicator, Electron Microscopy Sciences
Catalog # 100493-162
Supplier:  
Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Desiccator Cabinet, Bronze
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm (1/4") acrylic with full-length hinges and stainless steel twist locking latches.
  • Easy viewing clear units or dark bonze for light sensitive materials
  • Deep, slide-in trays with perforated bottom
  • Available in three sizes
Spécifications
  • Désignation:
    Low Temperature Desiccator Cabinet, Bronze