Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder
Catalog # 100491-988
Supplier #:  72645
Microbeaker Holder, Electron Microscopy Sciences
Catalog # 100491-988
Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder, Electron Microscopy Sciences
Microbeaker Holder
Price
$134.96
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Brushed aluminum rack holds six 12 mm O.D. microbeakers in unique, stepped "see-through" holes for illuminated and microscopic observation of samples.
Specifications
  • Désignation:
    Microbeaker Holder