Scintillation Vial Rack, Electron Microscopy Sciences
Product Details & Documents
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
Spécifications
- Désignation:Scintilation vial rack, welded
- No. of Places:100
- W×D×H:14½×15×2½"