Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Plain slides, 1 g, 1.2 mm, 75×25 mm (3×1")
Catalog # 100491-390
Supplier #:  63711-05
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Catalog # 100491-390
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Plain slides, 1 g, 1.2 mm, 75×25 mm (3×1")
Price
$119.11
Log in for availability
Each Gross of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.
Specifications
  • Capacité:
    1 g
  • Désignation:
    Plain slides
  • L×l:
    75×25 mm (3×1")
  • Épaisseur:
    1.2 mm