DrainRack™ Slide Rack, Electron Microscopy Sciences
Blue
  100490-986
 :  71446-B
DrainRack™ Slide Rack, Electron Microscopy Sciences
  100490-986
DrainRack™ Slide Rack, Electron Microscopy Sciences
DrainRack™ Slide Rack, Electron Microscopy Sciences
Blue
98,25 $
Each Pack of 10
Made from high impact polystyrene. This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individual numbered slots.
All sides are stored upright for easy insertion and removal. A unique feature with the DrainRack™ is the ability to read bar codes without having to remove the slides from the rack. For space saving purposes, you can double the amount of slides simply by storing 2 slides per slot, giving you a capacity of 200 slides per tray instead of 100. The racks are not autoclavable. They measure: 75 x 231 x 25 mm (3 x 9 x 1").
  • Color:
    Blue