Disposable Glass Tubes, Electron Microscopy Sciences
Disposable Glass Tubes, Electron Microscopy Sciences
Orange Label, 100 mm, 13 mm
Catalog # 100490-618
Supplier #:  64636-O
Disposable Glass Tubes, Electron Microscopy Sciences
Catalog # 100490-618
Disposable Glass Tubes, Electron Microscopy Sciences
Disposable Glass Tubes, Electron Microscopy Sciences
Orange Label, 100 mm, 13 mm
Price
$549.01
Log in pour la disponibilité
Each Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tubes are made from borosilicate glass, round bottom and plain top.
The tubes measuring 16 mm in outer diameter and 125 mm in length are Flint glass only.
Labeling area in five pastel colors. Handy fused-in color patches, accept any type of ink or pencil.
Spécifications
  • Ø:
    13 mm
  • Désignation:
    Orange Label
  • Hauteur:
    100 mm