Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
- Ø int.:45 mm
- Capacité:30 Dram (111 ml)
- Hauteur:76 mm