Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Product Details & Documents
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
Spécifications
- Ø int.:22 mm
- Capacité:5 Dram (18.5 ml)
- Hauteur:40 mm