Polystyrene Containers, Electron Microscopy Sciences
Product Details & Documents
These clear and rigid containers are made from polystyrene.
- All containers are aseptically manufactured
- Labeling options include unlabeled, printed, or plain
- 30mL containers with universal closure rely on special seal technology
- Other sizes employ liners or crab claw closures to facilitate sample containment
- Some sizes are available with spoons
- For in vitro use only
Spécifications
- Cap Type:PE, plain cap
- Ø:44.6 mm
- Capacité:60 ml
- Désignation:Container
- Hauteur:61.1 mm
- Label Type:Printed