Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grid slides, poly-l-lysine
  100490-302
 :  63406-02
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
  100490-302
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Grid slides, poly-l-lysine
$482.79
Each Pack of 144
These 3×1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents.
  • Each square grid fills one low-powered-field (approximately 100× magnification)
  • Easy to locate an organism or cell every time
  • Eighty one squares
The slides are available with coatings of Poly-L-Lysine or Silane, or they can be purchased plain (uncoated). The grid measures 2 mm square, 16 mm total. Numbers 1-8 and alpha A-H.
  • Désignation:
    Grid slides, poly-l-lysine