Emery Paper, Electron Microscopy Sciences | Avantor
Emery Paper, Electron Microscopy Sciences
Emery paper, 1 grit, 280 mesh
Catalog # 100490-136
Fournisseur #:  62117-10
Emery Paper, Electron Microscopy Sciences
Catalog # 100490-136
Emery Paper, Electron Microscopy Sciences
Emery Paper, Electron Microscopy Sciences
Emery paper, 1 grit, 280 mesh
Price
$116.40
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This emery paper comes in sheets and in varying grit sizes.
Grit 4/0 is the finest and 3 is the coarsest.
Size: 22×34 cm (9×13¾")
Spécifications
  • Désignation:
    Emery paper, 1 grit, 280 mesh