Ampoule Cracker, Electron Microscopy Sciences | Avantor
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Breaker, 10 - 20 ml ampoules
Catalog # 100490-064
Fournisseur #:  60605-20
Ampoule Cracker, Electron Microscopy Sciences
Catalog # 100490-064
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Breaker, 10 - 20 ml ampoules
Price
$301.16
Log in pour la disponibilité
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Ampoule Cracker™ is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules.
Simply slip the Ampoule Cracker™ over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
Spécifications
  • Désignation:
    Ampoule Breaker
  • For:
    10 - 20 ml ampoules