Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Product Details & Documents
Aluminum Oxide and Silicon Carbide Abrasive Cut-Off Wheels are offered for long life, and are affordably priced for various sectioning requirements.
All wheels are 228.6 mm (9") in diameter, 1.6 mm (0.062”) thick, and with 31 mm (1 1/4”) arbor.
Specifications
- Désignation:Soft Steel Rc30-45 Cut-Off Wheel