Dispensing Bottles, Electron Microscopy Sciences | Avantor
Dispensing Bottles, Electron Microscopy Sciences
150 ml (5 oz.)
Catalog # 100489-772
Fournisseur #:  64294
Dispensing Bottles, Electron Microscopy Sciences
Catalog # 100489-772
Dispensing Bottles, Electron Microscopy Sciences
Dispensing Bottles, Electron Microscopy Sciences
150 ml (5 oz.)
Price
$65.69
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Low density polyethylene bottles.
  • Tip has a 0.76 mm (0.003") opening
By squeezing the bottle a steady stream of liquid is produced.
Spécifications
  • Capacité:
    150 ml (5 oz.)