Cryo Labels, Electron Microscopy Sciences | Avantor
Cryo Labels, Electron Microscopy Sciences
Cryo labels, 25×50 mm, 200 labels/pack
Catalog # 100489-746
Minority or Woman-Owned Business Enterprise
Fournisseur #:  62046
Cryo Labels, Electron Microscopy Sciences
Catalog # 100489-746
Minority or Woman-Owned Business Enterprise
Cryo Labels, Electron Microscopy Sciences
Cryo Labels, Electron Microscopy Sciences
Cryo labels, 25×50 mm, 200 labels/pack
Price
$253.40
Log in pour la disponibilité
Each Pack of 200
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A special formulated material for use at ultra-low temperatures and in cryo storage conditions, such as liquid nitrogen and oxygen.
  • Will not peel or shrink
They are durable and adhere well to plastic as well as cardboard cryogenic storage boxes. Ten sheets of labels: five preprinted, five plain.
 : Packaged as 20 labels/sheet.
Spécifications
  • Désignation:
    Cryo labels
  • Dimensions:
    25×50 mm
  • Packaging:
    200 labels/pack
  • Cat. No.:
    100489-746