Microscope Slide, Frosted End, Electron Microscopy Sciences | Avantor
Microscope Slide, Frosted End, Electron Microscopy Sciences
Glass slides frosted end, 1 g
Catalog # 100489-400
Fournisseur #:  71870-20
Microscope Slide, Frosted End, Electron Microscopy Sciences
Catalog # 100489-400
Microscope Slide, Frosted End, Electron Microscopy Sciences
Microscope Slide, Frosted End, Electron Microscopy Sciences
Glass slides frosted end, 1 g
Price
$2,296.55
Log in pour la disponibilité
Each Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These slides have ¾" sandblasted frosting on one end which allows for easy marking.
Spécifications
  • Capacité:
    1 g
  • Désignation:
    Glass slides frosted end