Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences | Avantor
Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
Model 1
Catalog # 100489-340
Fournisseur #:  71548-08
Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
Catalog # 100489-340
Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
Model 1
Price
$332.20
Log in pour la disponibilité
Each Pack of 200
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Holds up to 5 slides 3x1"in size . These mailers are rigid, strong and durable with a convenient flip-top, snap-on lid.
When it stands in its upright position, it can be used as a staining container, with a minimal amount of solution being used. Packaged 25 mailers/pk, 8 packs/cs.
Spécifications
  • Désignation:
    Model 1