Microscope Slide Folder, Electron Microscopy Sciences
Yellow
Catalog # 100489-320
 :  71502-Y10
Microscope Slide Folder, Electron Microscopy Sciences
Catalog # 100489-320
Microscope Slide Folder, Electron Microscopy Sciences
Microscope Slide Folder, Electron Microscopy Sciences
Yellow
Price
336,97 $
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from heat resistant injection molded plastic. The Slide Folder™ will hold up to twenty standard microscope slides 75 x 25 mm (3x1").
The Slide Folder™ is made of two parts: a base holding the slides horizontally offering numbered space for easy identification, and transparent doors which can either cover the slides or be swung behind the SlideFolder™ for space conservation while using the tray. The base is available in 5 different colors, offering color-coding for each job. Each slide location is identified with a number from 1 to 20. Removal of the slides is made easy by simply pressing on one end, which will lift the other end up. The two transparent hinged covers offer a full view of each stored slide, even bar codes can be read without removing of the slide. All units are stackable and will resist temperatures between –80°C and +100°C. They are not autoclavable.
Spécifications
  • Color:
    Yellow