Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Product Details & Documents
Made from translucent polypropylene.
- Resistant to breakage
- Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
Spécifications
- Capacité:2 ml
- Désignation:Graduated