PTFE Printed Slides, Electron Microscopy Sciences | Avantor
PTFE Printed Slides, Electron Microscopy Sciences
Hydrophobic printed slides, 8 well, 6 mm
Catalog # 100488-900
Fournisseur #:  63422-06
PTFE Printed Slides, Electron Microscopy Sciences
Catalog # 100488-900
PTFE Printed Slides, Electron Microscopy Sciences
PTFE Printed Slides, Electron Microscopy Sciences
Hydrophobic printed slides, 8 well, 6 mm
Price
$335.89
Log in pour la disponibilité
Each Pack of 72
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These slides are extremely hydrophobic, acetone proof, chemical resistant and autoclavable. They are perfect for controlling cross-contamination in staining, immunolabeling and immunofluorescence tests.
  • Super hydrophobic mask
  • Clean, wettable wells
  • A bioadhesive surface treatment to enhance cell and tissue attachment
The well surface is 100% wettable, which enhances cell attachment and increases its spreading capabilities within the well. These slides minimize the use of expensive purified antigens and antibodies. The slides measure 3 x 1" (76 x 26 x 1 mm).
Manufactured to order offering maximum shelf life. The shelf life of 18 months to two years. However, it is recommended that all glass slides (Printed or Un-printed) be used in a timely manner to avoid glass "weathering", (slides becoming cloudy or hazy even with proper storage. All printed slides are autoclavable and have a frosted end. They are intended for one time use only.
Spécifications
  • Désignation:
    Hydrophobic printed slides, 8 well
  • L×l:
    6 mm