Syringes Disposable, Electron Microscopy Sciences
These high quality disposable rubber piston syringes offer substantial savings.
- Lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out
- Syringes are ETO gas sterilized, latex and pyrogen free
- Bold graduations are easily read and resist wipe-off
: All syringes are packaged in individual sterile strip packs.
- Capacité:2.5 ml
- Tip Style:Luerslip Centric