CoverWell™ Imaging Chambers, Electron Microscopy Sciences
Product Details & Documents
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Spécifications
- Profondeur:2.8 mm
- Désignation:1 well, 750 µl
- Taille:20 mm dia