Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi-Temp Indicator Tape, ¾" Wide
Catalog # 100488-166
Supplier #:  77015-07
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Catalog # 100488-166
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi Temperature Indicator Tape, Electron Microscopy Sciences
Hi-Temp Indicator Tape, ¾" Wide
Price
$211.55
Log in pour la disponibilité
Each Roll of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specialty cloth-textured Hi Temperature Indicator tape shows the word “STERILIZED” in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550°F dry oven temperatures. Available in widths of 1/2", 3/4", and 1" on 250” rolls.
Spécifications
  • Désignation:
    Hi-Temp Indicator Tape, ¾" Wide