Disposable Pipettes – Sterile, Electron Microscopy Sciences
Product Details & Documents
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Spécifications
- Volume:1 ml
- Packed:Single Wrap