4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Product Details & Documents
Biopsy processing cassettes with four square compartments each measuring 13 mm.
- Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Spécifications
- Color:Peach