4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Orange
Catalog # 101413-218
Supplier #:  70075-O
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Catalog # 101413-218
Supplier:  
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Orange
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Biopsy processing cassettes with four square compartments each measuring 13 mm.
  • Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Specifications
  • Color:
    Orange