Loop and Needle - Disposable, Electron Microscopy Sciences | Avantor
Loop and Needle - Disposable, Electron Microscopy Sciences
Loop, 10 µl, Yellow
Catalog # 101412-652
Fournisseur #:  62450-50
Loop and Needle - Disposable, Electron Microscopy Sciences
Catalog # 101412-652
Loop and Needle - Disposable, Electron Microscopy Sciences
Loop and Needle - Disposable, Electron Microscopy Sciences
Loop, 10 µl, Yellow
Price
$267.88
Log in pour la disponibilité
Each Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from Polystyrene.
Spécifications
  • Volume:
    10 µl
  • Color:
    Yellow
  • Désignation:
    Loop