Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences
2% Formaldehyde/0.02% Picric Acid in 0.1 M Sodium Phosphate Buffer (pH 7.2)
  100503-954
Supplier #:  15956
Fixatives, Prepared, Electron Microscopy Sciences
  100503-954
 :  
Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences
2% Formaldehyde/0.02% Picric Acid in 0.1 M Sodium Phosphate Buffer (pH 7.2)
 :  
118,59 $
Each (10ml)
Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.
Certifications: Certificates of analysis are available.
  • Désignation:
    2% Formaldehyde/0.02% Picric Acid in 0.1 M Sodium Phosphate Buffer (pH 7.2)