Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Dichloromethane Wash Bottle, 500 ml (16.9 oz.)
  100503-268
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
  100503-268
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Dichloromethane Wash Bottle, 500 ml (16.9 oz.)
$35.57
Each
Permanently labeled/color-coded wash bottles.
Pack of six - Variety includes: Acetone, Methanol, Water, Iso-propyl Alcohol, Ethanol and Toluene. Toluene Wash Bottle is for short term storage only.
These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz. (500 ml), low density polyethylene.
  • Capacité:
    500 ml (16.9 oz.)
  • Désignation:
    Dichloromethane Wash Bottle