Stabilizing Rings, Electron Microscopy Sciences | Avantor
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, 48 mm id
Catalog # 100501-586
Fournisseur #:  64436
Stabilizing Rings, Electron Microscopy Sciences
Catalog # 100501-586
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, 48 mm id
Price
$93.89
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Spécifications
  • Désignation:
    Stabilizing Ring, 48 mm id