Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Bowl size 28×65 mm, 30 cm
Catalog # 100501-466
Supplier #:  72984-30
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Catalog # 100501-466
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Bowl size 28×65 mm, 30 cm
Price
$208.74
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Ellipso-Spoon® Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Its unique shape means no more material loss. This utensil’s specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.
Specifications
  • Désignation:
    Bowl size 28×65 mm
  • Longueur:
    30 cm