BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
Product Details & Documents
Available in two specially shaped capsules which are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. The bottle neck capsule produces a 7.9 mm OD block with a hemihyperpoloid (rather than faced) tip. The conical capsule produces a 7.9 mm OD block with a conical (rather than faced) tip.
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Spécifications
- Désignation:Embedding Capsule Conical Tip