pH Indicator Papers - Test Papers, Electron Microscopy Sciences
pH Indicator Papers - Test Papers, Electron Microscopy Sciences
0.2/0.4, 7.9 - 9.8
Catalog # 100501-050
Supplier #:  65510-60
pH Indicator Papers - Test Papers, Electron Microscopy Sciences
Catalog # 100501-050
pH Indicator Papers - Test Papers, Electron Microscopy Sciences
pH Indicator Papers - Test Papers, Electron Microscopy Sciences
0.2/0.4, 7.9 - 9.8
Price
$68.93
Log in for availability
Box of 100 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
In contrast to conventional indicator papers, the indicator dyes of pH-Fix sticks are substantially bound to the cellulose fibers
Measurement of pH value is possible even in unbuffered or very weakly buffered solutions, since the sticks can be left in the solution until the final color change is completed. Test samples are not contaminated by the indicator dyes. Colors of the individual color fields cannot run into each other, thereby allowing for a more precise comparison with the color scale. Recently developed indicator dyes guarantee a sharp differentiation between the individual pH values and a clear and easy comparison to the color scale. Sample application - four different indicator papers 6x5 mm are sealed on to a plastic strip 6 x 85 mm. The strips are sufficiently long to avoid contact between the fingers and the test sample.
As a result, the possibility of the color bleeding, even in strongly basic solutions, is avoided.
Specifications
  • pH Range:
    7.9 - 9.8
  • pH Graduation:
    0.2/0.4