Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
50 ml, Sample Vials with Blue Caps, Non sterile
Catalog # 100500-420
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Catalog # 100500-420
Supplier:  
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
50 ml, Sample Vials with Blue Caps, Non sterile
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
TUBE CENTRIFUGE ATTACH RD CAP 50ML CS500 Catalog #76452-254
Product 76452-254 is similar to 100500-420 but may have a different manufacturer and/or packaging.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transport vials/tubes are ideal for sample collection and storage.
  • Conical tubes with freestanding skirts
  • Tubes are autoclavable
  • Write-on area on side
Transport Vials are made from virgin polypropylene. They are available in three sizes, and come either sterile or non sterile. Graduations provide convenient and accurate references.
Transport Tubes are made from virgin polypropylene (PP) or polystyrene (PS). They also work well in Olympus automated testing equipment. Plug caps are required.
Delivery information: 5 ml and 10 ml vials are supplied with white screw cap and 50 mL vials are supplied with blue cap that has a molded-in sealing ring for a leak-proof fit. Plug caps are available in yellow color and you can choose from hollow top (HT) or hollow bottom (HB).
Spécifications
  • Sterility:
    Non sterile
  • Volume:
    50 ml
  • Désignation:
    Sample Vials with Blue Caps
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Hollow Bottom Plug Cap, Yellow
    Hollow Bottom Plug Cap, Yellow
    100490-282
    Log in pour la disponibilité
    Each - $104.86
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Hollow Top Plug Cap, Yellow
    Hollow Top Plug Cap, Yellow
    100490-284
    Log in pour la disponibilité
    Each - $104.86