Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences | Avantor
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
Catalog # 100499-706
Fournisseur #:  71156
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Catalog # 100499-706
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
Price
425,88 $
Log in pour la disponibilité
Dozen Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
  • Made from a special plastic that minimizes static
  • Complete with grid recording card
Spécifications
  • Capacité:
    100 Grids
  • Désignation:
    Grid Storage Box
  • Dimensions:
    8 L×5 W×7 T mm