Workstation Storage Dispenser Bins, Electron Microscopy Sciences | Avantor
Workstation Storage Dispenser Bins, Electron Microscopy Sciences
Storage/Dispenser Bin, 6x6.5x7
Catalog # 100498-688
Fournisseur #:  64364-07
Workstation Storage Dispenser Bins, Electron Microscopy Sciences
Catalog # 100498-688
Workstation Storage Dispenser Bins, Electron Microscopy Sciences
Storage/Dispenser Bin, 6x6.5x7
Price
$382.69
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Workstation storage dispenser bins fabricated from white and clear acrylic.
  • Hinged top cover assures easy filling
  • Hinged front cover provides a generous opening for easy removal
  • All items that are stored remain dust-free, yet transparent through the clear front window
  • Available in two sizes
Spécifications
  • Désignation:
    Storage/Dispenser Bin, 6x6.5x7