Bright-Line Counting Chamber, Electron Microscopy Sciences
Bright-Line Counting Chamber, Electron Microscopy Sciences
White Cell Pipette (Thoma) (1458)
  100498-508
Supplier #:  63514-32
Bright-Line Counting Chamber, Electron Microscopy Sciences
  100498-508
Bright-Line Counting Chamber, Electron Microscopy Sciences
Bright-Line Counting Chamber, Electron Microscopy Sciences
White Cell Pipette (Thoma) (1458)
$173.64
Each
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
Delivery information: The complete set includes one counting chamber, two 0.4 mm cover glass, one red cell pipette, one white cell pipette, tubing, mouthpieces, and storage case.
  • Désignation:
    White Cell Pipette (Thoma) (1458)