Diamond Points, Electron Microscopy Sciences
Diamond Points, Square Grid, 3.8 mm, Cone
  100497-982
 :  62155-38
Diamond Points, Electron Microscopy Sciences
  100497-982
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Square Grid, 3.8 mm, Cone
$65.77
Each
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
  • Ø:
    3.8 mm
  • Désignation:
    Diamond Points, Square Grid
  • Shape:
    Cone