Hex Key Wrenches, Electron Microscopy Sciences
Product Details & Documents
Made from heat treated chrome nickel steel. Useful to have around the lab. Its sturdy handle, folds up just like a pocket knife. Can be used at any angle.
Specifications
- Désignation:Hex Key, Metric Set 2
- Cat. No.:100497-570
- Type:Hex Key