Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks (Stainless Steel), 1.25"
Catalog # 100496-974
 :  50152-03
Flat Mounting Blocks , Electron Microscopy Sciences
Catalog # 100496-974
Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks (Stainless Steel), 1.25"
Price
169,24 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.
Spécifications
  • Ø:
    1.25"
  • Désignation:
    Flat Mounting Blocks (Stainless Steel)