Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Clear-Mount , Electron Microscopy Sciences

Clear-Mount , Electron Microscopy Sciences

Clear-Mount, (with Tris Buffer), 250 ml
Catalog # 100496-548
Supplier #:  17985-15
Clear-Mount , Electron Microscopy Sciences
Catalog # 100496-548
CAS Number:  
Clear-Mount , Electron Microscopy Sciences
Clear-Mount , Electron Microscopy Sciences

Clear-Mount, (with Tris Buffer), 250 ml

CAS Number:  
Price
$726.35
Log in for availability
Each (250ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Application: Mounting of Immunohisto slides

Clear-Mount is a water based mounting medium designed for the permanent mounting of hydrated tissues, which may be damaged with organic solvents.  Such samples include cell smears, with peroxidase and alkaline phosphatase chromogens. This mounting medium preserves Fast Red, Aminoethylycarbazole (AEC), NBT/BCIP, INT/BCIP chromogens and is also compatible with counter-stain such as Hematoxylin and Nuclear Fast Red. It is also suitable for chromogens like DAB and DAB with Nickel and Cobalt. It is not compatible with H and E staining.

Application: Mounting of Immunohisto slides. Avalaible in original fomula with Tris Buffer or with PIPE Buffer for a more stable pH.

*The names BioMeda™ Gel Mount and Crystal Mount are all owned by BioMeda Corporation

*The names BioMeda™ Gel Mount and Crystal Mount are all owned by BioMeda Corporation

Avalaible in original fomula with Tris Buffer or with PIPE Buffer for a more stable pH.

Specifications

Specifications

  • Désignation:
    Clear-Mount, (with Tris Buffer)
  • Taille:
    250 ml

Specifications

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.