Element Standard Sample Storage Box, Electron Microscopy Sciences | Avantor
Element Standard Sample Storage Box, Electron Microscopy Sciences
Element Standard Storage Box, 20 Discs
Catalog # 100495-006
Fournisseur #:  80063-20
Element Standard Sample Storage Box, Electron Microscopy Sciences
Catalog # 100495-006
Element Standard Sample Storage Box, Electron Microscopy Sciences
Element Standard Storage Box, 20 Discs
Price
389,55 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
It is designed to store the element discs securely, therefore, avoiding any contact with some of the dangerous elements.
A sample storage box that holds either 10 or 20 of ¼" diameter discs.
Spécifications
  • Désignation:
    Element Standard Storage Box, 20 Discs