MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Featherlite Vannas Scissors Straight
  100494-436
 :  77900-S
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
  100494-436
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Featherlite Vannas Scissors Straight
$772.64
Each
Featherlite scissors in different styles.
Style DS-Straight Vannas Scissors, 5 mm and smooth blades. Length 125 mm (5").
Style DS-G – Stork Style, 10 mm long and smooth blades. Length: 125 mm (5").
Style DS-GM – Micro Stork Style, 5 mm long and smooth blades. Length: 125 mm (5").
  • Désignation:
    Featherlite Vannas Scissors Straight