Thin Sample Holder, Electron Microscopy Sciences
Thin sample holder, 8 mm Pin
  100494-354
 :  75948-08
Thin Sample Holder, Electron Microscopy Sciences
  100494-354
Thin Sample Holder, Electron Microscopy Sciences
Thin Sample Holder, Electron Microscopy Sciences
Thin sample holder, 8 mm Pin
$130.41
Each
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals and more.
  • Désignation:
    Thin sample holder, 8 mm Pin