Dissecting Scissors - Operating Scissors, Electron Microscopy Sciences
Product Details & Documents
They come in a variety of styles and lengths:
Made from stainless steel.
They come in a variety of styles and lengths:
S/B - One sharp point and one blunt point
S/S - Two sharp points
B/B - Two blunt points
Specifications
- Désignation:Straight
- Longueur:5½"
- Blade Type:Sharp/Sharp