Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Product Details & Documents
All Vannas micro scissors have extra fine tips and are 3" (75 mm) long. Available with a 3 mm or 5 mm cutting edge.
Spécifications
- Désignation:Vannas Scissors Curved, Delicate
- Blade Length:3 mm (⅛")
- Blade Type:Extra fine