Vial Rack, Electron Microscopy Sciences | Avantor
Vial Rack, Electron Microscopy Sciences
Rack for 1 Dram Shell Vial, 94 x 266 x 28 mm (3 3/4 x 10 5/8 x 1 1/8")
Catalog # 100493-822
Fournisseur #:  72641-01
Vial Rack, Electron Microscopy Sciences
Catalog # 100493-822
Vial Rack, Electron Microscopy Sciences
Vial Rack, Electron Microscopy Sciences
Rack for 1 Dram Shell Vial, 94 x 266 x 28 mm (3 3/4 x 10 5/8 x 1 1/8")
Price
$150.22
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This rugged polypropylene rack holds 48 vials, size 1 dram (15 x 45 mm); or 4 ml tissue culture vials; or 2 mL serum vials in 4 x 12 configuration with alpha-numeric indexing.
The rack is constructed from chemically resistant, autoclavable (121 °C for 20 minutes) polypropylene and features corner posts to assure stability when stacking empty.
Spécifications
  • Désignation:
    Rack for 1 Dram Shell Vial
  • Dimensions:
    94 x 266 x 28 mm (3 3/4 x 10 5/8 x 1 1/8")